missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ HumanKine™ Human Thermostable bFGF Recombinant Protein, Xeno-Free

Supplier:  MilliporeSigma™ GF44650UG

 View more versions of this product

Catalog No. GF44650UG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Human Thermostable bFGF Recombinant Protein, Xeno-Free
Human 293 protein expression system
Endotoxin free and greater than 95% pure by SDS-PAGE
50 μg
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions