missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ HPTLC LiChrospher Silica Gel 60 Plates/Sheets

thin layer chromatography plates based on spherical silica particles

$1008.30 - $1433.66

Specifications

Analysis Time 3 to 20 min.
Pore Size 60 Å
Excitation Wavelength 254 nm
For Use With (Application) Fast Analysis of Complex Samples for Manual or Instrumental Use
Pore Volume 0.1 to 0.5 μL
View More Specs

Products 3
Catalog Number Mfr. No. Dimensions (L x W) Thickness (Metric) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Thickness (Metric) Price Quantity  
M1055860001
View Documents
MilliporeSigma™
1055860001
20 x 20 cm 200 μm
Each for $1,168.82
Only null left
 
M1154450001
View Documents
MilliporeSigma™
1154450001
20 x 10 cm 200 μm
Each for $1,433.66
Only null left
 
M1056470001
View Documents
MilliporeSigma™
1056470001
20 x 10 cm 100 μm
Each for $1,008.30
Only null left
 
Description

Description

Unique HPTLC Lichrospher™ plates are the first thin layer chromatography plates based on spherical silica particles. They offer the ultimate in thin layer chromatography performance and speed enabling high throughput analysis of complex samples.
Specifications

Specifications

3 to 20 min.
254 nm
0.1 to 0.5 μL
20 cm
60 Å
Fast Analysis of Complex Samples for Manual or Instrumental Use
Silica Gel 60 F254 s
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'