Learn More
MilliporeSigma™ HPTLC LiChrospher Silica Gel 60 Plates/Sheets
thin layer chromatography plates based on spherical silica particles
Supplier: MilliporeSigma™ 1055860001
Description
Unique HPTLC Lichrospher™ plates are the first thin layer chromatography plates based on spherical silica particles. They offer the ultimate in thin layer chromatography performance and speed enabling high throughput analysis of complex samples.Specifications
| 3 to 20 min. | |
| 60 Å | |
| Silica LiChrospher HPTLC Plate | |
| Fast Analysis of Complex Samples for Manual or Instrumental Use | |
| Silica Gel 60 F254 s | |
| 20 x 20 cm | |
| 20 cm |
| Aluminum | |
| 25 Sheets/Pk. | |
| 254 nm | |
| 0.1 to 0.5 μL | |
| 200 μm | |
| 20 cm |