Learn More
MilliporeSigma™ HPTLC LiChrospher Silica Gel 60 Plates/Sheets
thin layer chromatography plates based on spherical silica particles
Supplier:  MilliporeSigma™ 1056470001
 View more versions of this product
Catalog No. M1056470001
Only null left
Explore available promotions
Description
Description
Unique HPTLC Lichrospher™ plates are the first thin layer chromatography plates based on spherical silica particles. They offer the ultimate in thin layer chromatography performance and speed enabling high throughput analysis of complex samples.
Specifications
Specifications
3 to 20 min.
25 Plates/Pk.
254 nm
0.1 to 0.5 μL
100 μm
20 cm
60 Å
Silica LiChrospher HPTLC Plate, Extra Thin
Fast Analysis of Complex Samples for Manual or Instrumental Use
Silica Gel 60 F254 s
20 x 10 cm
10 cm
Product Suggestions
Product Suggestions