missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Enzyme Free Cell Dissociation Solution Hank's Based (1x)

Hank's based formulation of chelating agents and agents to stabilize their activity on cells

$31.30 - $75.80

Specifications

Content And Storage 4°C
Formulation Chelating and stabilizing agents
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S004C
View Documents
MilliporeSigma™
S004C
100 mL
Each for $31.30
Only null left
 
S004B
View Documents
MilliporeSigma™
S004B
500 mL
Each for $75.80
Only null left
 
Description

Description

Enzyme-Free Cell Dissociation Solution is a Hank's based formulation of chelating agents and agents to stabilize their activity on cells. This reagent does NOT contain porcine trypsin or any other protein. It works significantly faster than 0.5mM EDTA. In fact, it works as quickly as trypsin on most cells. Most importantly, it preserves the structural and functional integrity of cell surface proteins and does not have the cytotoxic effects sometimes associated with 0.5mm EDTA. It can be used to dissociate primary cells, tissues, and tumors with an increased plating efficiency.
Specifications

Specifications

4°C
Chelating and stabilizing agents
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'