missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EmbryoMax™ 2X Cell Culture Freezing Medium

Formulated with 20% DMSO and Fetal Bovine Serum

$268.00 - $463.00

Specifications

Content And Storage -80°C up to 2 years
For Use With (Application) Stem cell culture
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ES00210F
View Documents Promotion Details
MilliporeSigma™
ES00210F
100 mL
Each for $463.00
Only null left
 
ES002D
View Documents Promotion Details
MilliporeSigma™
ES002D
50 mL
Each for $268.00
Only null left
 
Description

Description

This unique formulation of cell culture freezing media is for cryopreservation of a broad spectrum of mammalian cells (see viability data). Freezing Media contain 10% of the active cryopreservation ingredient (either DMSO or Glycerol). The remaining 90% is a proprietary mixture of Calf and Fetal Bovine Serum. The reagents used in these products are certified to be virus and mycoplasma free.
Specifications

Specifications

-80°C up to 2 years
Stem cell culture
Videos
SDS
Documents

Documents