missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EmbryoMax™ 2X Cell Culture Freezing Medium

Catalog No. ES002D
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES002D 50 mL
ES00210F 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES002D Supplier MilliporeSigma™ Supplier No. ES002D

Formulated with 20% DMSO and Fetal Bovine Serum

This unique formulation of cell culture freezing media is for cryopreservation of a broad spectrum of mammalian cells (see viability data). Freezing Media contain 10% of the active cryopreservation ingredient (either DMSO or Glycerol). The remaining 90% is a proprietary mixture of Calf and Fetal Bovine Serum. The reagents used in these products are certified to be virus and mycoplasma free.

Specifications

Content And Storage -80°C up to 2 years
For Use With (Application) Stem cell culture
Quantity 50 mL