Learn More
MilliporeSigma™ Chromolith™ SemiPrep Guard Cartridges
Catalog No. M1520350001
Change view
Click to view available options
Sorbent:
SemiPrep RP-18 endcapped sorbent
SemiPrep Si sorbent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sorbent
M1520350001 SemiPrep Si sorbent
M1520360001 SemiPrep RP-18 endcapped sorbent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1520350001 Supplier MilliporeSigma™ Supplier No. 1.52035.0001
For prolonging the lifespan of Chromolith™ analytical columns
Although the Chromolith™ columns are well known for their robustness and longevity, guard columns are available to prolong the lifespan of Chromolith™ analytical columns even further. Package includes three guard cartridges.
Specifications
Quantity 1/Pk.
Length (Metric) 10 mm
Type HPLC Guard Cartridge
Diameter (Metric) Inner 10 mm
Sorbent SemiPrep Si sorbent