Learn More
MilliporeSigma™ Chromolith™ RP Endcapped HPLC Guard Cartridges
Also popular for the protection of classical packed HPLC columns
Supplier: MilliporeSigma™ 1520090001
Description
- Handles easily
- Adds to the robustness and longevity of monolithic analytical columns
Specifications
| 3/Pk. | |
| HPLC Guard Cartridge | |
| RP-18 endcapped sorbent |
| 5 mm | |
| 2 mm |