Learn More
MilliporeSigma™ Chromolith™ HighResolution RP-18 Endcapped HPLC Guard Cartridge
For prolonging the lifespan of Chromolith™ analytical columns
Supplier: MilliporeSigma™ 1520250001
Description
Although the Chromolith™ columns are well known for their robustness and longevity, guard columns are available to prolong the lifespan of Chromolith™ analytical columns even further. These guard cartridges require a special holder.Specifications
| 1/Pk. | |
| 5 mm | |
| HPLC Cartridge Holder | |
| 4.6 mm |