Learn More
MilliporeSigma™ Chemicon™ Recombinant Mouse Leukemia Inhibitory Factor
biologically active and suitable for cell culture applications.
Supplier:  MilliporeSigma™ LIF2050
Catalog No. LIF2050
Only null left
Description
Description
Leukaemia inhibitory factor is a cytokine that induces macrophage differentiation. Neurotransmitters and neuropeptides, well known for their role in the communication between neurons, are also capable of activating monocytes and macrophages and inducing chemotaxis in immune cells. LIF signals through different receptors and transcription factors. LIF in conjunction with BMP2 acts in synergy on primary fetal neural progenitor cells to induce astrocytes.
Specifications
Specifications
2- 8°C for 6 - 12 months
Cell culture
50 μg
E. coli
Product Suggestions
Product Suggestions