Learn More
MilliporeSigma™ Chemicon™ Recombinant Human Leukemia Inhibitory Factor
Biologically active and suitable for cell culture applications.
Supplier:  MilliporeSigma™ LIF1050
Catalog No. LIF1050
Only null left
Explore available promotions
Description
Description
Leukemia inhibitory factor is a cytokine that induces macrophage differentiation. Neurotransmitters and neuropeptides, well known for their role in the communication between neurons, are also capable of activating monocytes and macrophages and inducing chemotaxis in immune cells. LIF signals through different receptors and transcription factors. LIF in conjunction with BMP2 acts in synergy on primary fetal neural progenitor cells to induce astrocytes.
Specifications
Specifications
2- 8°C for 6 - 12 months
Liquid
Greater than 95% by SDS-PAGE. Endotoxin level is less than 0.1 ng per mg of LIF. Tested negative in both aseptic and mycoplasmic tests.
E. coli
100 μg/mL
Cell culture, stem cell culture
50 μg
Product Suggestions
Product Suggestions