missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ MitoLight™ Mitochondrial Apoptosis Detection Kit

For flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker

Specifications

Content And Storage -20°C
Description Mitochondrial Apoptosis Detection Kit
Detection Method Fluorescence
For Use With (Application) For flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker
Includes 10X Incubation Buffer, MitoLight
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
APT242
View Documents Promotion Details
MilliporeSigma™
APT242
100 Tests N/A
Only null left
 
APT142
View Documents Promotion Details
MilliporeSigma™
APT142
25 Tests N/A
Only null left
 
Description

Description

Specifications

Specifications

-20°C
Fluorescence
10X Incubation Buffer, MitoLight
Mitochondrial Apoptosis Detection Kit
For flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker
Videos
SDS
Documents

Documents