Learn More
MilliporeSigma™ Chemicon™ MitoLight™ Mitochondrial Apoptosis Detection Kit
For flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker
Supplier:  MilliporeSigma™ APT142
 View more versions of this product
Catalog No. APT142
Only null left
Description
Description
Specifications
Chemical Identifiers
-20°C
Fluorescence
10X Incubation Buffer, MitoLight
Mitochondrial Apoptosis Detection Kit
For flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker
Specifications
-20°C
Fluorescence
10X Incubation Buffer, MitoLight
Mitochondrial Apoptosis Detection Kit
For flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker
25 Tests
Product Suggestions
Product Suggestions