missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ PEG 6000

Catalog No. 528877100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
528877100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 528877100GM Supplier MilliporeSigma™ Supplier No. 528877100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular Biology Grade

PEG 6000, Molecular Biology Grade - CAS 25322-68-3 - Calbiochem

Specifications

Content And Storage 15°C to 30°C
Product Type PEG 6000
Quantity 100 g