missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Molecular Biology Grade Sucrose

CAS 57-50-1

$782.00 - $782.00

Specifications

Content And Storage 15-30°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5731135KG
View Documents
MilliporeSigma™
5731135KG
5kg
Each for $782.00
Only null left
 
Description

Description

Specifications

Specifications

15-30°C
Videos
SDS
Documents

Documents