Learn More
MilliporeSigma™ Calbiochem™ Human Plasma alpha1-Acid Glycoprotein
Native α1-acid glycoprotein from human plasma used for the study of the structure of oligosaccharide units
Supplier:  MilliporeSigma™ 1122511MG
Catalog No. 1122511MG
Only null left
Explore available promotions
Description
Description
. Consists of 40 - 45% carbohydrate. Present in human plasma at concentrations of 55 - 140mg/100 ml. Clinically, α1-AG is an acute-phase reactant that, together with haptoglobin, indicates acute inflammation. Selectively suppresses the augmentation of NK cell activity by IFN-α and IFN-γ. The α1-AG:α1-antitrypsin ratio is useful in diagnosis of liver disease. A sialic acid-deficient α1-AG has been observed in serum or urine of patients with Hodgkin’s disease, psoriatic arthritis, diabetes, and chronic myeloid leukemia. Sialic acid-deficient α1-AG has an affinity for vitamin B12.
Specifications
Specifications
2-8°C
1 mg
Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
Product Suggestions
Product Suggestions