missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ CELLULYSIN™ Cellulase

For isolating intact cells and protoplasts from plant tissues

$181.04 - $666.51

Specifications

Content And Storage 2-8°C
For Use With (Application) Isolate intact cells and protoplasts from plant tissues
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
21946650KU
View Documents
MilliporeSigma™
21946650KU
50 KU N/A
Only null left
 
219466100KU
View Documents
MilliporeSigma™
219466100KU
100 KU N/A
Only null left
 
219466200KU
View Documents
MilliporeSigma™
219466200KU
200 KU N/A
Only null left
 
Description

Description

Hydrolyzes β1,4-glucan linkages in cellulose. Has been used to isolate intact cells and protoplasts from plant tissues.
Specifications

Specifications

2-8°C
Isolate intact cells and protoplasts from plant tissues
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'