Learn More
MilliporeSigma™ Amicon™ Ultra Vials
Catalog No. UFC50VL96
Change view
Click to view available options
Volume (Metric):
0.5 mL
Catalog No. Volume (Metric)
UFC50VL96 0.5 mL
1 options
Catalog No. UFC50VL96 Supplier MilliporeSigma™ Supplier No. UFC50VL96
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessory for Amicon Ultra-0.5 centrifugal filters
  • Volume: 0.5 mL
  • For protein purification, western blotting, protein extraction
Specifications
Disposable Single-use
Sterility Non-sterile
For Use With (Application) Protein purification, protein extraction, protein concentration and desalting, western blotting
No. per Pack 96
Quantity 96Pack
Type Centrifugal Filters
Volume (Metric) 0.5 mL