missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Amicon™ Pro Purification System

Catalog No. acs500024
Change view
Click to view available options
Quantity:
24/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACS500024 24/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACS500024 Supplier MilliporeSigma™ Supplier No. ACS500024
Only null left

Collapses the protein purification workflow into as few as three spins, in a single device

This part number excludes Amicon Ultra 0.5 filter.

Only one device required for sample prep workflow, including:

  • Purification
  • Concentration
  • Desalting/buffer exchange
Superior sample recovery:
  • No need to transfer
  • Better protein binding with packed agarose resin compared to batch mixing
Ergonomic, less pipetting:
  • No need to transfer
  • Simultaneous concentration capability:
    • Elution without dilution
    • Consistent recovery
    • Saves time
    Large reservoir saves time:
    • Holds wide range of volumes
    • Reduces spin steps
    • Reservoir cap can be labeled to reduce time and confusion
    Gentle and efficient buffer exchange:
    • >99.99% (1000-fold) desalting in single spin
    • Maintains protein activity
    • Broad reagent compatibility
    • Agarose resin kits optimized for the Amicon™ Pro device
    • Compatible with other resins >10μm in diameter
    • Chemically compatible with most common buffers

    Specifications

    Includes 24 Exchange devices, and 24 Collection tubes with cap
    Quantity 24/Pk.
    Type Pro Affinity Purification System
    Length (English) 4.7 in.
    Length (Metric) 12 cm