missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Protein G Reagent Kit
Reagents for Amicon Pro Affinity Concentration Kit Protein G with Amicon Ultra–0.5 Device
Supplier: MilliporeSigma™ ACR5000PG
Description
For use with the Amicon Pro Affinity Conc: 3 ml Protein G resin, 2 X 66ml Bind/Wash Buffer, 12 ml Elution buffer, 2.5 ml Neutralization bufferSpecifications
| Single-use | |
| Non-sterile | |
| Regenerated Cellulose (RC) | |
| 1Pack | |
| 15 mL |
| Amicon Pro Affinity | |
| Antibody purification | |
| 3 mL Protein G resin, 2 x 66 mL Bind/Wash Buffer, 12 mL Elution buffer, 2.5 mL Neutralization buffer | |
| Centrifugal Filters |