Learn More
MilliporeSigma™ Amicon™ Pro Affinity concentration Kit Ni–NTA with Amicon™ Ultra–0.5 Device: Reagents
Reagents for MilliporeSigma™ Amicon Pro Affinity Concentration Kit Ni–NTA with Amicon Ultra–0.5 Device
Supplier: MilliporeSigma™ ACR5000NT
Description
For use with EMD Millipore Amicon Pro Affinity Concentration Kit Ni–NTA with Amicon Ultra–0.5 Device (Cat. No. ACK5030NT)
- 3mL HisBind Ni–NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose)
- 25mL Bind Buffer (4X)
- 25mL Wash buffer (4X)
- 5mL elution buffer (4X)
Specifications
| Amicon Pro Affinity | |
| 3mL HisBind Ni-NTA resin, 25mL Bind Buffer (4x), 25mL Wash buffer (4x), 5mL elution buffer (4x) | |
| Ni-NTA Reagent Kit |
| His-tagged protein purification | |
| 1/Pk. |