Learn More
MilliporeSigma™ Amicon™ Pro Affinity concentration Kit GST with Amicon™ Ultra–0.5 Device
GST Reagent Kit for GSH-tagged protein purification.
Supplier: MilliporeSigma™ ACR5000GS
Description
For use with the Amicon Pro Affinity Conc: 3mL GST Bind resin, 125 mg Glutathione (reduced, free acid), 25ml GST Bind/Wash Buffer (10X), 5mL GST Reconstitution buffer (10X)Specifications
| Amicon Pro Affinity | |
| 3mL GST Bind resin, 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) | |
| GST Reagent Kit |
| GSH-tagged protein purification | |
| 1/Pk. |