Learn More
MilliporeSigma™ Alizarin-Red Staining Solution
a commonly used stain to identify calcium containing osteocytes in differentiated culture of both human and rodent mesenchymal stem cells (MSCs).
Supplier:  MilliporeSigma™ TMS008C
Catalog No. TMS008C
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Stem cell culture, cell based assays, cell differentiation
4.1±0.1
2 x 50 mL
Product Suggestions
Product Suggestions