missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermus thermophilus DNA Ligase

Catalog No. FEREL07981
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREL07981 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREL07981 Supplier Thermo Scientific™ Supplier No. EL07981

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Thermus thermophilus DNA Ligase is a thermostable NAD-dependent DNA ligase that catalizes formation of phosphodiester bonds between 5'-phospate and 3'-hydroxyl termini in double-stranded DNA.

Thermo Scientific™ Thermus thermophilus DNA Ligase is a thermostable NAD-dependent DNA ligase that catalizes formation of phosphodiester bonds between 5`-phospate and 3`-hydroxyl termini in double-stranded DNA. Thermus thermophilus DNA Ligase is not active on RNA, single-stranded DNA, or blunt-ended DNA.

Features

  • Increased specificity—high ligation temperature eliminates non-specific ligation
  • Stabile at high temperatures with an optimum ligation temperature of 45°C to 65°C

Applications

Thermus thermophilus DNA ligase is recommended for applications requiring high ligation specificity, including:

  • Single-stranded nick repair in double-stranded DNA
  • Next-generation sequencing
  • Rolling circle amplification
  • Proximity ligation assay

Specifications

Concentration 50 U/μL
For Use With (Application) Single-stranded nick repair, NGS, RCA, PLA
Content And Storage 10X Thermus thermophilus DNA Ligase Buffer, 40 μL

Store at -20°C.
Enzyme DNA Ligase
Sensitive to Heat Inactivation No
Quantity 1000 U
Product Type NAD-dependent DNA Ligase
Research Category Next-generation Sequencing, DNA Repair
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.