Learn More
Ellagic acid hydrate, 97%, may cont. up to 12% water
CAS: 476-66-4 | C14H6O8 | 302.19 g/mol
Supplier: Thermo Scientific Chemicals A1572214
Description
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEllagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.
Solubility
Slightly soluble in water. Soluble in 1M sodium hydroxide (10 mg/ml), EtOH, and pyridine.
Notes
Air sensitive and light sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container. Store in dark and away form air.
Specifications
| Ellagic acid hydrate | |
| 450°C (decomposition) | |
| C14H6O8 | |
| 25 g | |
| Air and light sensitive | |
| ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique | |
| AFSDNFLWKVMVRB-UHFFFAOYSA-N | |
| 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0â´,¹â¶.0¹¹,¹âµ]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione | |
| 5281855 | |
| 302.20 (Anhydrous) |
| 476-66-4 | |
| may cont. up to 12% water | |
| MFCD00006914 | |
| 47549 | |
| 14,3547 | |
| Slightly soluble in water. Soluble in 1M sodium hydroxide (10mg/ml),EtOH,and pyridine. | |
| OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O | |
| 302.19 | |
| CHEBI:4775 | |
| 97% |
Chemical Identifiers
| 476-66-4 | |
| 302.19 | |
| AFSDNFLWKVMVRB-UHFFFAOYSA-N | |
| 5281855 | |
| 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0â´,¹â¶.0¹¹,¹âµ]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione |
| C14H6O8 | |
| MFCD00006914 | |
| ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique | |
| CHEBI:4775 | |
| OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-508-3
RTECSNumber : DJ2620000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only