accessibility menu, dialog, popup

UserName

CAS RN 476-66-4

IUPAC Name: 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
Synonyms: ellagic acid|acid, ellagic|acido elagico|alizarine yellow|acide ellagique|gallogen|ellagic acid dihydrate|ellagic acid hydrate|ellagate|benzoaric acid
Molecular Weight (g/mol): 302.19
Molecular Formula: C14H6O8
InChi Key: AFSDNFLWKVMVRB-UHFFFAOYSA-N
SMILES: OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)