missing translation for 'onlineSavingsMsg'
Learn More

Ellagic Acid Dihydrate 98.0+%, TCI America™

Catalog No. p-7128968
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03755G 5 g
E037525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03755G Supplier TCI America Supplier No. E03755G
Only null left

Chemical Identifiers

CAS 133039-73-3
Molecular Formula C14H10O10
Molecular Weight (g/mol) 338.22
MDL Number MFCD00149494
InChI Key ZEPCRIPMALGRJR-UHFFFAOYSA-N
Synonym 2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione
PubChem CID 16760409
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4,6,8(16),11,13-hexaene-3,10-dione dihydrate
SMILES O.O.OC1=C(O)C2=C3C(=C1)C(=O)OC1=C3C(=CC(O)=C1O)C(=O)O2

Specifications

Melting Point 450°C
Quantity 5 g
Formula Weight 302.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ellagic Acid Dihydrate
RTECSNumber DJ2620000
TSCA No