accessibility menu, dialog, popup

UserName

CAS RN 133039-73-3

IUPAC Name: 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4,6,8(16),11,13-hexaene-3,10-dione dihydrate
Synonyms: ellagic acid dihydrate
Molecular Weight (g/mol): 338.22
Molecular Formula: C14H10O10
InChi Key: ZEPCRIPMALGRJR-UHFFFAOYSA-N
SMILES: O.O.OC1=C(O)C2=C3C(=C1)C(=O)OC1=C3C(=CC(O)=C1O)C(=O)O2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results