missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ellagic acid, 97%

CAS: 476-66-4 | C14H6O8 | 302.19 g/mol

Supplier:  Thermo Scientific Chemicals 117740100

 View more versions of this product

Catalog No. AC117740100

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ellagic acid
>360°C
Glass bottle
MFCD00006914
19, 261
ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
AFSDNFLWKVMVRB-UHFFFAOYSA-N
6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
5281855
302.19
476-66-4
97%
C14H6O8
10 g
15, 3602
Solubility in water: slightly soluble. Other solubilities: soluble in alkalies and pyridine,practically insoluble in ether
OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O
302.19
CHEBI:4775
97%

Chemical Identifiers

476-66-4
302.19
AFSDNFLWKVMVRB-UHFFFAOYSA-N
5281855
6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
C14H6O8
MFCD00006914
ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
CHEBI:4775
OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

EINECSNumber : 207-508-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only