missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ellagic acid, 97%

Catalog No. AC117740025
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117740025 2.5 g Glass bottle
AC117740100 10 g Glass bottle
AC117740500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117740025 Supplier Thermo Scientific Chemicals Supplier No. 117740025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 476-66-4 | C14H6O8 | 302.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Melting Point >360°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 2.5 g
Beilstein 19, 261
Merck Index 15, 3602
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkalies and pyridine,practically insoluble in ether
Formula Weight 302.19
Percent Purity 97%
Chemical Name or Material Ellagic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 207-508-3

RUO – Research Use Only