missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA-Light™ Immunoassay System with CDP-Star™ and Sapphire-II™ Substrate/Enhancer Solution

Catalog No. T1025
Encompass
Change view
Click to view available options
Quantity:
1,000 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1025 1,000 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1025 Supplier Invitrogen™ Supplier No. T1025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ultra-sensitive detection of antigens or antibodies in multiple immunoassay formats employing an alkaline phosphatase enzyme label.

  • Heterogeneous assay format increases sensitivity and dynamic range and reduces interference by test compounds in the detection portion of the assay (as compared with homogeneous assay formats)
  • Chemiluminescent immunoassay detection allows greater sensitivity and improved dynamic range as compared to radioimmunoassays and immunoassays that employ colorimetric or fluorescent detection
  • High sensitivity and wide dynamic range permit sensitive detection with fewer assays, because sample dilution to fit the dynamic range of the assay is unnecessary
  • A variety of ready-to-use substrate formulations lets the user precisely optimize signal intensity to the exact requirements of an assay
  • The kinetics of the glow-assay provides a window during which measurements may be performed, facilitating HTS applications where assay automation is used
  • Chemiluminescent immunoassay detection provides versatile applications such as sandwich immunoassays, competitive immunoassays, and DNA probe capture assays
  • Non-radioactive immunoassays eliminate concerns over use of radioisotopes

Extend the Limits of Detection
The ELISA-Light system offers an excellent combination of high sensitivity and throughput for assays measured in a luminometer. The ELISA-Light system significantly extends the limits of detection of most immunoassays previously performed with colorimetric and isotopic methods.

Multiple Substrate/Enhancer Formulations
The kit contains reagents optimized for chemiluminescent ELISAs employing alkaline phosphatase labels and is available with a choice of ready-to-use chemiluminescent substrate formulations containing CSPD or CDP-Star™ substrate with Sapphire-II or Emerald-II™ luminescence enhancers. Multiple substrate/enhancer formulations provide options that let the user optimize specific assays or luminometers. The specific formulation that will provide the highest sensitivity for a specific assay can vary. Therefore, an ELISA sampler kit containing all four substrate/enhancer formulations is available to facilitate assay development.

Specifications

  • Detection Method: Chemiluminescent
  • For Use With (Equipment): Microplate Reader
  • Label or Dye: AP Alkaline Phosphatase

Specifications

Product Type Immunoassay System
Quantity 1,000 assays
Detection Method Chemiluminescence
Conjugate AP
For Use With (Equipment) Microplate Reader
Product Line ELISA-Light
Substrate Type Alkaline Phosphatase
Content And Storage This kit contains enough reagent for 1000 assays. Store at 4°C.
What is the typical format of an ELISA kit and what is included in each kit?

All ELISA kits are provided in the sandwich ELISA format with capture antibody already coated onto a 96 well plate. Typical detection uses a biotinylated detection antibody followed by Streptavidin-HRP and HRP substrate. Most kits are available as single 96-well plate kits, some are available as 2- and 5-plate kits. Kits typically contain:

96 Well Strip Plate coated with capture antibody
Standard protein
Wash buffer, 10x
ELISA buffer/Diluent, 10x
Detection antibody (in most kits, biotinylated)
HRP Reagent (either secondary antibody or streptavidin conjugated to HRP)(100x)
Substrate (usually TMB)
Stop solution
Adhesive plate covers

Should I run any internal controls with your ELISA kits? Where can I get internal controls?

Running internal controls along with the blanks and the standard curve is an excellent way to monitor the performance of any ELISA kit. Internal controls are samples containing known amounts of the analyte being measured with the kit. A key attribute of internal controls, and what differentiates them from the samples of the standard curve, is that they duplicate the composition of your actual samples as closely as possible. This is important because your samples may contain components that affect detection of the analyte differently than does the Standard Diluent provided in the kit. In other words, internal controls allow you to determine if assaying the same amount of analyte in the Standard Diluent (the standard curve) and in the sample matrix (internal controls) gives the same results. It's also important to remember that internal controls give you confidence in the accuracy of your results. Internal controls help ensure that the assay is performing consistently from assay to assay, every time you run it.

One source of internal controls is naturally occurring samples (such as serum or plasma) containing known amounts of the analyte you want to measure. Such positive control samples can be run as-is, or they can be diluted to various known concentrations, preferably with a naturally occurring negative sample of the same type. It's a good idea to run at least 2 internal controls, if you can. One usually has an analyte concentration between the lowest point on the standard curve and the curve's midpoint, while the other has a concentration between the midpoint and the highest concentration. Some researchers also run an internal control that falls close to the midpoint of the curve.

If you don't have access to naturally occurring controls, you can prepare them yourself. Let's say that you will be measuring interleukin-6 (IL-6) in human serum samples with an ELISA kit, Cat. No. KHC0061. First you will need to obtain some pooled normal human sera, which will be used as the sample matrix for your internal controls. Preferably, the IL-6 content of this serum should be negligible (but known) or too low to measure. Next you should add known amounts of human IL-6 to this serum to create the internal controls, using the human IL-6 standard provided in the kit. After you prepare the standard curve as described in the product manual, add some of the leftover concentrated IL-6 standard to the serum matrix. As described above, you can prepare controls with high and low IL-6 levels, or high, medium, and low, if you have enough wells in your ELISA plate.

Even if your sample type is not serum and you're not measuring IL-6, follow the general procedure outlined above to prepare your internal controls with your protein and sample matrix of interest. A valuable resource describing Spike and Recovery and Linearity of Dilution assays can be found here (http://tools.thermofisher.com/content/sfs/brochures/TR0058-Spike-and-Recovery.pdf). If you need more help, please contact Technical Support at techsupport@thermofisher.com.

What are the different types of ELISA kits you offer?

Our ELISA kits can be categorized into several different groups (see Table 2, Page 35 of the Protein Analysis Handbook - http://www.thermofisher.com/us/en/home/global/forms/protein-handbook-registration.html), based on a number of factors: target protein class, sensitivity, readout method, or ability to detect specific phosphorylation states of the target protein.

Standard colorimetric ELISA kits use a standard colorimetric readout and allow excellent sensitivity and detection range. Typical sensitivity is less than 10 pg/mL and standard curve range is around 10-250 pg/mL, although there are some kits with wider ranges.
Ultrasensitive ELISA kits use a standard colorimetric readout but enable detection and analysis of proteins to levels as low as 0.5 pg/mL. With measurement range of 0.5-20 pg/mL, these kits are especially useful with highly diluted samples.
Chemi ELISA kits use chemiluminescence detection for high sensitivity (less than 1 pg/mL) and are highly flexible with a wide measurement range from 0.5 to 2000 pg/mL.
Phospho ELISA kits enable the specific detection of phosphorylation of key signaling proteins with high specificity, and are often used to supplement western blot results and provide quantitative data.

What options do you offer for performing ELISAs?

We offer full ELISA kits, Reagent Sets and Antibody Pairs. ELISA kits are complete, ready-to-use kits with pre-coated plates, all buffers, capture, and detection antibodies included. Most kits are single plate format, but some are available in 2- or 5-plate formats. Reagent Sets are for researchers who need the core kit components but prefer to make their own buffers and coat their own plates. Antibody Pairs are matched pairs of detection and capture antibodies for researchers who need to process large numbers of samples.

For Research Use Only. Not for use in diagnostic procedures.