missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ELISA/ELISPOT Diluent (5X)

Catalog No. 501129752
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501129752 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501129752 Supplier Invitrogen™ Supplier No. 00420256

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supplemented with fetal bovine serum as source of animal serum proteins. Used for blocking non-specific binding of ELISA plates, diluting recombinant standards and test samples and making the working dilutions of secondary detection reagent. Also contains 1% ProClin 150 as a preservative

The 5X ELISA/ELISPOT Diluent is a Phosphate Buffered Solution supplemented with fetal bovine serum as a source of animal serum proteins. This buffer is used for blocking non-specific binding of ELISA plates, diluting recombinant standards and test samples, and making the working dilutions of the secondary detection reagent (i.e., detection antibody-biotin and AV-HRP Cat. No. 18-4100). This diluent solution also contains a preservative.

Reported Application
ELISA

Specifications

Product Type Diluent
Quantity 100 mL
Form Liquid
For Use With (Application) ELISA
Product Line eBioscience

For Research Use Only