missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical
Cleaning solution
Specifications
| Color | Pale Yellow |
|---|---|
| For Use With (Equipment) | Electrode |
| Type | Cleaning Solution |
| For Use With (Application) | Removal of membrane/junction Protein deposits |
| Packaging | Natural Poly Bottle |
Description
- Pale yellow liquid
- CAS: 7647-01-0 (Hydrochloric Acid); 9001-75-6 (Pepsin); 7732-18-5 (Water, Deionized)
Specifications
| Pale Yellow | |
| Cleaning Solution | |
| Natural Poly Bottle |
| Electrode | |
| Removal of membrane/junction Protein deposits |
Safety and Handling
ShelfLife : 1 Year