missing translation for 'onlineSavingsMsg'
Learn More

Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical

Catalog No. 279416
Change view
Click to view available options
Capacity (Metric):
20 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
279416 500 mL
279420 20 mL
279432 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 279416 Supplier Ricca Chemical Company Supplier No. 279416
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleaning solution

  • Pale yellow liquid
  • CAS: 7647-01-0 (Hydrochloric Acid); 9001-75-6 (Pepsin); 7732-18-5 (Water, Deionized)

Specifications

Capacity (English) 16.9 oz.
Capacity (Metric) 500 mL
Color Pale Yellow
For Use With (Equipment) Electrode
Type Cleaning Solution
For Use With (Application) Removal of membrane/junction Protein deposits
Packaging Natural Poly Bottle
Quantity 500 mL