missing translation for 'onlineSavingsMsg'
Learn More

Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical

Cleaning solution

Supplier:  Ricca Chemical Company 279420

 View more versions of this product

Catalog No. 279420


Only null left

Description

Description

  • Pale yellow liquid
  • CAS: 7647-01-0 (Hydrochloric Acid); 9001-75-6 (Pepsin); 7732-18-5 (Water, Deionized)

Specifications

Chemical Identifiers

Pale Yellow
Cleaning Solution
Natural Poly Bottle
Electrode
Removal of membrane/junction Protein deposits

Specifications

0.6 oz.
Pale Yellow
Cleaning Solution
Natural Poly Bottle
20 mL
Electrode
Removal of membrane/junction Protein deposits
20 x 20 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 1 Year

SDS
Documents

Documents

Promotions

Promotions