missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Elastase Substrate V

Catalog No. AAJ60093MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60093MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60093MB Supplier Thermo Scientific Chemicals Supplier No. J60093MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

627.70 g/mol

Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color White
Form Powder
For Use With (Application) Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase
Quantity 25 mg
Substrate Type Elastase Substrate
Molecular Weight (g/mol) 627.70
Storage Requirements -30° to -10°C
Substrate Quantity 25mg

RUO – Research Use Only