Learn More
Agilent BioTek EL406 Microplate Washer/Dispenser, IVD

The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek 406PUB2SI
Description
The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers. The EL406 capabilities include washing and dispensing for standard assays, plus biomagnetic separation and vacuum filtration protocols.
The Dual-Action manifold optimizes washing for loosely adherent cell monolayers. The patented Ultrasonic Advantage prevents blockages in the manifold tubes before washing, to reduce assay failure. This modular plate washer dispenser offers flexibility with both syringe pump and peristaltic pump dispensers, along with plate washing, on a single platform.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Chemical Identifiers
| ±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL | |
| Optional (1 or 2 L) | |
| 500 nL to 3000 μL/well | |
| ≤2 μL/well | |
| Microplate Washer/Dispenser | |
| 1 to 250 | |
| 50/60 Hz | |
| 16.5 in. | |
| 12.5 in. | |
| 18 in. | |
| 32 lb. |
| CE and TUV marked. RoHS compliant. | |
| Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL | |
| 96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates | |
| Programmable in minutes and seconds, up to 60 min. | |
| 14.5 kg | |
| High to Low Flow; Optimized rates for cell assays | |
| 1 μL | |
| 42 cm | |
| 31.8 cm | |
| 46 cm | |
| 100/240 V |
Specifications
| ±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL | |
| Optional (1 or 2 L) | |
| 500 nL to 3000 μL/well | |
| ≤2 μL/well | |
| Microplate Washer/Dispenser | |
| 1 to 250 | |
| 50/60 Hz | |
| 1 μL | |
| 16.5 in. | |
| 12.5 in. | |
| 18 in. | |
| 32 lb. |
| CE and TUV marked. RoHS compliant. | |
| Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL | |
| 96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates | |
| Programmable in minutes and seconds, up to 60 min. | |
| 14.5 kg | |
| High to Low Flow; Optimized rates for cell assays | |
| 406PUB2 | |
| Faster 384-well microplate washing (192-tube (16 x 12) manifold/steel tubes); 1-Reagent Dispense Module – 1 Peristaltic Pump (autoclavable, 8-tip (1 x 8) cassette/sapphire jeweled, steel or plastic tips); Ultrasonic Advantage; Automatic 4-Buffer Switching | |
| 42 cm | |
| 31.8 cm | |
| 46 cm | |
| 100/240 V |