missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EK-Away™ Resin
Specifically designed for removal of EKMaxor other enterokinase enzymes following cleavage of proteins containing the enterokinase cleavage site
$1352.00 - $1352.00
Specifications
| Content And Storage | EK-Away™ is provided as a 50% ethanol slurry and should be stored at +4°C. 10X binding and 10X stripping buffers are included. 7.5 ml resin will remove 250 units of enterokinase; 30 ml will remove 1000 units of EKMax™. All reagents are guaranteed stable for 6 months when stored properly. |
|---|---|
| Form | Liquid Suspension |
| Product Line | EK-Away |
| Type | Resin |
| Stationary Phase | Enterokinase |
Description
- The resin is conjugated with soybean trypsin inhibitor, which has a high affinity and binding capacity for enterokinase
- The enzyme catalytic site binds this agarose-based resin for simple batch or column-bound removal of EKMaxor other enterokinase preparations
GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
| EK-Away™ is provided as a 50% ethanol slurry and should be stored at +4°C. 10X binding and 10X stripping buffers are included. 7.5 ml resin will remove 250 units of enterokinase; 30 ml will remove 1000 units of EKMax™. All reagents are guaranteed stable for 6 months when stored properly. | |
| EK-Away | |
| Enterokinase |
| Liquid Suspension | |
| Resin | |
| Affinity |
For Research Use Only. Not for use in diagnostic procedures.