missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Efavirenz

Catalog No. AC466620050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466620050 5 g
AC466620250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466620050 Supplier Thermo Scientific Chemicals Supplier No. 466620050

CAS: 154598-52-4 | C14H9ClF3NO2, C14H9ClF3NO2 | 315.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154598-52-4
Molecular Formula C14H9ClF3NO2

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: insoluble.
Melting Point 138.0°C
Formula Weight 315.68
InChI Key XPOQHMRABVBWPR-ZDUSSCGKSA-N
SMILES FC(F)(F)[C@]1(OC(=O)NC2=CC=C(Cl)C=C12)C#CC1CC1
IUPAC Name (4S)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-2,4-dihydro-1H-3,1-benzoxazin-2-one
Molecular Weight (g/mol) 315.68
Chemical Name or Material Efavirenz
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only