missing translation for 'onlineSavingsMsg'
Learn More

EDTA Titrant, 0.01M solution, Macron Fine Chemicals™

macron

$395.56 - $4645.89

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MKH1051
View Documents
Avantor Macron
H10505
1 L
Each for $395.56
Case of 6 Each for $2,373.36
Only null left
 
MKH1054
View Documents
Avantor Macron
H10507
4 L
Case of 4 Each for $4,645.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

60-00-4
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
C10H16N2O8
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Colorless
C10H16N2O8
860mg/mL
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
6049
292.24
Crystals
60-00-4
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
292.24
CHEBI:42191
1 L
EDTA Titrant, 0.01M solution
Videos
SDS
Documents

Documents