missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Titrant, 0.0100 M (M/100), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 98.0, 0.36 |
| CAS Max % | 100.0, 0.38 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | 4 to 6 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.0100Molar |
| Chemical Name or Material | EDTA Titrant |