missing translation for 'onlineSavingsMsg'
Learn More

EDTA Titrant, 0.0100 M (M/100), Ricca Chemical

Catalog No. 27001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
27001 4 L Natural Poly Bottle
270016 500 mL Natural Poly Bottle
270032 1 L Natural Poly Bottle
R27000004F 4 L Cubitainer
270025 10 L Cubitainer
27005 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 27001 Supplier Ricca Chemical Company Supplier No. 27001
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6381-92-6 , 7732-18-5
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
CAS Min % 98.0, 0.36
CAS Max % 100.0, 0.38
Grade Laboratory
Packaging Natural Poly Bottle
pH 4 to 6
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.0100Molar
Chemical Name or Material EDTA Titrant