missing translation for 'onlineSavingsMsg'
Learn More

EDTA Standard Solution, 1mL = 1mg CaCO3, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$3289.56 - $4834.44

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02002794
View Documents
Avantor J.T.Baker
564802
1 L Narrow Mouth Polyethylene Bottle
Case of 6 Each for $3,289.56
Only null left
 
02002796
View Documents
Avantor J.T.Baker
564803
4 L Cubetainer
Case of 4 Each for $4,834.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
C10H18N2Na2O10
MFCD00150037,MFCD00003541
44120005
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Narrow Mouth Polyethylene Bottle
1L = 1.00kg
MFCD00150037,MFCD00003541
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
44120005
BAKER ANALYZED™ Reagent
6381-92-6
C10H18N2Na2O10
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
1 L
372.24
EDTA Standard Solution, 1mL = 1mg CaCO3
Videos
SDS
Documents

Documents