missing translation for 'onlineSavingsMsg'
Learn More

EDTA Standard Solution, 1mL = 1mg CaCO3, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002796
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002796 4 L Cubetainer
02002794 1 L Narrow Mouth Polyethylene Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002796 Supplier Avantor Supplier No. 564803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Packaging Cubetainer
Specific Gravity 1L = 1.00kg
Quantity 4 L
Formula Weight 372.24
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material EDTA Standard Solution, 1mL = 1mg CaCO3