missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Disodium, 0.1 M Solution, Spectrum™ Chemical
$179.03 - $2622.09
Specifications
| Chemical Name or Material | EDTA Disodium |
|---|---|
| CAS | 6381-92-6,7732-18-5 |
| CAS Max % | 3.7,96.3 |
| Concentration | 0.1M |
| Grade | Chemical Solution |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18610341
|
Spectrum Chemical Mfg Cor
E120500MLPL |
500 mL | Cubitainer |
Each for $179.03
|
|
||||
|
18610340
|
Spectrum Chemical Mfg Cor
E1204LTPL |
4 L | Cubitainer |
Each for $627.38
|
|
||||
|
18610339
|
Spectrum Chemical Mfg Cor
E12020LTBL |
20 L | Cubitainer |
Each for $2,622.09
|
|
||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.