missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium, 0.1 M Solution, Spectrum™ Chemical

Numéro de catalogue. 18610341
Modifier l'affichage
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Cubitainer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18610341 500 mL Cubitainer
18610340 4 L Cubitainer
18610339 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 18610341 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur E120500MLPL
Il en reste null

Spécifications

Chemical Name or Material EDTA Disodium
CAS 6381-92-6, 7732-18-5
CAS Max % 3.7, 96.3
Concentration 0.1M
Grade Chemical Solution
Packaging Cubitainer
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.