missing translation for 'onlineSavingsMsg'
Learn More

EDTA, 0.0100 Molar, Reagents

Catalog No. 01335359
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335359 10 L
01335358 1 L
01335357 20 L
01335356 4 L
01335355 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01335359 Supplier Labchem Inc Supplier No. CE25040010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EDTA, 0.0100 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 6381-92-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.002
pH 4 to 6
Quantity 10 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material EDTA, 0.0100 Molar