missing translation for 'onlineSavingsMsg'
Learn More

Econazole, 99%, Thermo Scientific Chemicals

Catalog No. AC458630250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458630250 25 g
AC458630050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458630250 Supplier Thermo Scientific Chemicals Supplier No. 458630250

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27220-47-9
Molecular Formula C18H15Cl3N2O
Molecular Weight (g/mol) 381.68
MDL Number MFCD00800993,MFCD00058160 (.HNO3)
InChI Key LEZWWPYKPKIXLL-UHFFFAOYNA-N
Synonym econazole, ecostatin, econazolum, pevaryl, gyno-pevaryl, econazol, palavale, spectazole, +--econazole, ecostatin cream
PubChem CID 3198
ChEBI CHEBI:82873
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole
SMILES ClC1=CC=C(COC(CN2C=CN=C2)C2=C(Cl)C=C(Cl)C=C2)C=C1

Specifications

Melting Point 91.0°C to 94.0°C
Loss on Drying 0.5% max. (1 g, 60°C, 4 h)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hot methanol, slightly soluble in ethanol, insoluble in ether
Formula Weight 381.68
Percent Purity 99%
Chemical Name or Material Econazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 248-341-6

RUO – Research Use Only