accessibility menu, dialog, popup

UserName

CAS RN 27220-47-9

IUPAC Name: 1-{2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl}-1H-imidazole
Synonyms: econazole econazole nitrate preparation econazole nitrate (substance) gynopevaril ecostatin vaginal ovules econazole (product) pevaryl spectazole econazole nitrate palavale
Molecular Weight (g/mol): 381.68
Molecular Formula: C18H15Cl3N2O
InChi Key: LEZWWPYKPKIXLL-UHFFFAOYNA-N
SMILES: ClC1=CC=C(COC(CN2C=CN=C2)C2=C(Cl)C=C(Cl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results